accessibility menu, dialog, popup

UserName

H3CNNOBr

CAS RN 56653-43-1

IUPAC Name: 2-bromo-1-(1-methyl-1H-1,3-benzodiazol-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(1-methyl-1,3-benzodiazol-2-yl)ethanone
Molecular Weight (g/mol): 253.1
Molecular Formula: C10H9BrN2O
InChi Key: KHFRWYRANOMZCP-UHFFFAOYSA-N
SMILES: CN1C(=NC2=CC=CC=C12)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results