CAS RN 56287-63-9
CAS RN 56287-63-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-Viloxazine Hydrochloride, TRC
CAS: 56287-63-9 Molecular Formula: C13H19NO3 • HCl Molecular Weight (g/mol): 237.293646 Synonym: (R)-2-[(2-Ethoxyphenoxy)methyl]morpholine Hydrochloride,(R)-2-(2-Ethoxyphenoxymethyl)tetrahydro-1,4-oxazine Hydrochloride IUPAC Name: (2R)-2-[(2-ethoxyphenoxy)methyl]morpholine;hydrochloride SMILES: CCOC1=CC=CC=C1OC[C@H]2CNCCO2.Cl