accessibility menu, dialog, popup

UserName

SOOClONHONH

CAS RN 56044-12-3

IUPAC Name: 2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride
Synonyms: 2,4-dioxo-1,3-dihydroquinazoline-6-sulfonyl chloride
Molecular Weight (g/mol): 260.65
Molecular Formula: C8H5ClN2O4S
InChi Key: LEFXQSDJQGOMPS-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C2NC(=O)NC(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results