CAS RN 5561-87-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 3-Hydroxydecanoic Acid, TRC
CAS: 5561-87-5 Molecular Formula: C10 H20 O3 Molecular Weight (g/mol): 188.26 IUPAC Name: 3-hydroxydecanoic acid SMILES: CCCCCCCC(O)CC(=O)O