CAS RN 55368-42-8
CAS RN 55368-42-8
IUPAC Name:
4-bromobenzene-1-carboximidamide hydrochloride
Synonyms:
4-bromobenzenecarboximidamide hydrochloride
Molecular Weight (g/mol):
235.51
Molecular Formula:
C7H8BrClN2
InChi Key:
IMTHEBSPHHMJOJ-UHFFFAOYSA-N
SMILES:
Cl.NC(=N)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
4-Bromobenzamidine hydrochloride, Thermo Scientific™
CAS: 55368-42-8 Molecular Formula: C7H8BrClN2 Molecular Weight (g/mol): 235.509 MDL Number: MFCD00464967 InChI Key: IMTHEBSPHHMJOJ-UHFFFAOYSA-N Synonym: 4-bromobenzamidine hydrochloride,4-bromobenzimidamide hydrochloride,4-bromobenzamidine hcl,4-bromo-benzamidine hydrochloride,4-bromobenzenecarboximidamide hydrochloride,4-bromobenzene-1-carboximidamide hydrochloride,benzenecarboximidamide, 4-bromo-, monohydrochloride,4-bromobenzenecarboxamidine, chloride,benzenecarboximidamide,4-bromo,4-bromobenzamidinehydrochloride PubChem CID: 12207714 IUPAC Name: 4-bromobenzenecarboximidamide;hydrochloride SMILES: C1=CC(=CC=C1C(=N)N)Br.Cl