accessibility menu, dialog, popup

UserName

OHOOO

CAS RN 552-86-3

IUPAC Name: 1,2-bis(furan-2-yl)-2-hydroxyethan-1-one
Synonyms: furoin
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: MIJRFWVFNKQQDK-UHFFFAOYNA-N
SMILES: OC(C(=O)C1=CC=CO1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results