accessibility menu, dialog, popup

UserName

CAS RN 54960-65-5

SOOOHNHHOHOHO

CAS RN 54960-65-5

IUPAC Name: 4-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}butane-1-sulfonic acid
Synonyms: 4-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}butane-1-sulfonic acid
Molecular Weight (g/mol): 257.3
Molecular Formula: C8H19NO6S
InChi Key: VFTZCDVTMZWNBF-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)NCCCCS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results