accessibility menu, dialog, popup

UserName

CAS RN 54770-27-3

BrBrPP(E)

CAS RN 54770-27-3

IUPAC Name: triphenyl[(1E)-2-(triphenylphosphaniumyl)ethenyl]phosphanium dibromide
Synonyms: triphenyl[(1E)-2-(triphenylphosphaniumyl)ethenyl]phosphanium dibromide
Molecular Weight (g/mol): 710.43
Molecular Formula: C38H32Br2P2
InChi Key: TVFYBHGFLHYJNV-GLDAUVFXSA-L
SMILES: [Br-].[Br-].C(=C/[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)\[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results