accessibility menu, dialog, popup

UserName

CAS RN 5472-41-3

NH2NONHNHN

CAS RN 5472-41-3

IUPAC Name: 4-amino-1H,2H,6H-pyrazolo[3,4-d]pyrimidin-6-one
Synonyms: 4-amino-1H,2H-pyrazolo[3,4-d]pyrimidin-6-one
Molecular Weight (g/mol): 151.13
Molecular Formula: C5H5N5O
InChi Key: KTQYLKORCCNJTQ-UHFFFAOYSA-N
SMILES: NC1=NC(=O)N=C2NNC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results