accessibility menu, dialog, popup

UserName

H3CH3CCH3CH3H3COHOH3CCH3H3C

CAS RN 54680-48-7

IUPAC Name: 2-(4,4-dimethylpentan-2-yl)-5,7,7-trimethyloctanoic acid
Synonyms: 2-(4,4-dimethylpentan-2-yl)-5,7,7-trimethyloctanoic acid
Molecular Weight (g/mol): 284.48
Molecular Formula: C18H36O2
InChi Key: QBTIHZIVENIGSW-UHFFFAOYNA-N
SMILES: CC(CCC(C(C)CC(C)(C)C)C(O)=O)CC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)