accessibility menu, dialog, popup

UserName

CAS RN 5464-79-9

CH3OSH2NN

CAS RN 5464-79-9

IUPAC Name: 4-methoxy-1,3-benzothiazol-2-amine
Synonyms: 4-methoxy-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 180.23
Molecular Formula: C8H8N2OS
InChi Key: YEBCRAVYUWNFQT-UHFFFAOYSA-N
SMILES: COC1=C2N=C(N)SC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)