accessibility menu, dialog, popup

UserName

FFFBrSOOCl

CAS RN 54403-98-4

IUPAC Name: 2-bromo-4-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 2-bromo-4-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 323.51
Molecular Formula: C7H3BrClF3O2S
InChi Key: WWEGTYZLWBOTQW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results