CAS RN 54322-32-6
CAS RN 54322-32-6
IUPAC Name:
ethyl N-(2,4-dinitrophenoxy)ethanimidate
Synonyms:
ethyl N-(2,4-dinitrophenoxy)ethanimidate
Molecular Weight (g/mol):
269.21
Molecular Formula:
C10H11N3O6
InChi Key:
UVBZLMGBNXCYNA-UHFFFAOYSA-N
SMILES:
CCOC(C)=NOC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O
1
–
1
of
1
results
1
Ethyl N-(2,4-dinitrophenoxy)acetimidate, 98+%, Thermo Scientific™
CAS: 54322-32-6 Molecular Formula: C10H11N3O6 Molecular Weight (g/mol): 269.213 MDL Number: MFCD00010133 InChI Key: UVBZLMGBNXCYNA-YRNVUSSQSA-N Synonym: ethyl n-2,4-dinitrophenoxy acetimidate,pubchem12897,jlyppahrtackn bt jw dtttrrvbapemaadmu@dmp,ethyl e-n-2,4-dinitrophenoxy acetimidate,1-1e-2-ethoxy-1-azaprop-1-enyloxy-2,4-dinitrobenzene PubChem CID: 9566065 IUPAC Name: ethyl (1E)-N-(2,4-dinitrophenoxy)ethanimidate SMILES: CCOC(=NOC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C