CAS RN 5421-46-5
CAS RN 5421-46-5
IUPAC Name:
ammonium 2-sulfanylacetate
Synonyms:
ammonium thioglycolate
Molecular Weight (g/mol):
109.14
Molecular Formula:
C2H7NO2S
InChi Key:
ZZTCCAPMZLDHFM-UHFFFAOYSA-N
SMILES:
[NH4+].[O-]C(=O)CS
1
–
1
of
1
results
1
Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5 | CAS: 5421-46-5 | C2H7NO2S | 109.143 g/mol