accessibility menu, dialog, popup

UserName

NaCH3CH3HOSOHOO

CAS RN 540-92-1

IUPAC Name: 2-hydroxypropane-2-sulfonic acid sodium
Synonyms: 2-hydroxypropane-2-sulfonic acid sodium
Molecular Weight (g/mol): 163.14
Molecular Formula: C3H8NaO4S
InChi Key: RJQNNQWGSIPDML-UHFFFAOYSA-N
SMILES: [Na].CC(C)(O)S(O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results