CAS RN 53985-49-2
IUPAC Name:
3-chloro-5-(trifluoromethyl)benzoic acid
Synonyms:
3-chloro-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
224.56
Molecular Formula:
C8H4ClF3O2
InChi Key:
IDLOGBMWOUINGG-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(Cl)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Chloro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 53985-49-2 Molecular Formula: C8H4ClF3O2 Molecular Weight (g/mol): 224.563 MDL Number: MFCD04115846 InChI Key: IDLOGBMWOUINGG-UHFFFAOYSA-N Synonym: 3-chloro-5-trifluoromethyl benzoic acid,3-chloro-5-trifluoromethyl benzoicacid,3-chloro-5-trifluoromethyl-benzoic acid,acmc-1axez,ksc495i8p,3-carboxy-5-chlorobenzotrifluoride,5-chloro-3-trifluoromethyl benzoic acid,benzoic acid, 3-chloro-5-trifluoromethyl,5-chloro-alpha,alpha,alpha-trifluoro-m-toluic acid PubChem CID: 3802362 IUPAC Name: 3-chloro-5-(trifluoromethyl)benzoic acid SMILES: C1=C(C=C(C=C1C(F)(F)F)Cl)C(=O)O