accessibility menu, dialog, popup

UserName

OOHN

CAS RN 53937-02-3

IUPAC Name: 4-(benzyloxy)-1,2-dihydropyridin-2-one
Synonyms: 4-(benzyloxy)-1H-pyridin-2-one
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: DOVNUEPFPBWTSV-UHFFFAOYSA-N
SMILES: O=C1NC=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results