CAS RN 53896-49-4
CAS RN 53896-49-4
IUPAC Name:
pyridazine-3-carbonitrile
Synonyms:
pyridazine-3-carbonitrile
Molecular Weight (g/mol):
105.1
Molecular Formula:
C5H3N3
InChi Key:
PJESVVYWPFAJCS-UHFFFAOYSA-N
SMILES:
N#CC1=NN=CC=C1
1
–
1
of
1
results
1
3-Cyanopyridazine, 97%
CAS: 53896-49-4 Molecular Formula: C5H3N3 Molecular Weight (g/mol): 105.1 MDL Number: MFCD09881239 InChI Key: PJESVVYWPFAJCS-UHFFFAOYSA-N Synonym: 3-cyanopyridazine,3-pyridazinecarbonitrile,3-cyano-1,2-diazine,pyridazine-3-carbonitrile PubChem CID: 13642940 IUPAC Name: pyridazine-3-carbonitrile SMILES: C1=CC(=NN=C1)C#N