accessibility menu, dialog, popup

UserName

OOHHN

CAS RN 5382-49-0

IUPAC Name: 1,2,3,4-tetrahydroquinoline-6-carboxylic acid
Synonyms: 1,2,3,4-tetrahydroquinoline-6-carboxylic acid
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: ARNALYPZOYPNAF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(NCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results