CAS RN 53081-25-7
CAS RN 53081-25-7
IUPAC Name:
(2R,3R,4S,5S,6R)-3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Synonyms:
(2R,3R,4S,5S,6R)-3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular Weight (g/mol):
540.66
Molecular Formula:
C34H36O6
InChi Key:
OGOMAWHSXRDAKZ-BWNLSPMZSA-N
SMILES:
O[C@@H]1O[C@H](COCC2=CC=CC=C2)[C@H](OCC2=CC=CC=C2)[C@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1
1
–
2
of
2
results
1
2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%
CAS: 53081-25-7 Molecular Formula: C34H36O6 MDL Number: MFCD00132927
2
2,3,4,6-Tetra-O-benzyl-D-galactopyranose, TRC
CAS: 53081-25-7 Molecular Formula: C34 H36 O6 Molecular Weight (g/mol): 540.65 IUPAC Name: (3R,4S,5S,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol SMILES: OC1O[C@H](COCc2ccccc2)[C@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@H]1OCc5ccccc5