CAS RN 53-94-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Hydroxy-2-aminofluorene, TRC
CAS: 53-94-1 Molecular Formula: C13H11NO Molecular Weight (g/mol): 197.23 Synonym: N-Hydroxy-9H-fluoren-2-amine,2-(Hydroxyamino)fluorene,N-2-Fluorenylhydroxylamine,N-Hydroxy-2-aminofluorene,N-Hydroxy-2-fluorenamine IUPAC Name: N-(9H-fluoren-2-yl)hydroxylamine SMILES: NC([C@@H](N)C([2H])([2H])[2H])=O.Cl