CAS RN 529-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Quinolin-4(1H)-one, TRC
CAS: 529-37-3 Molecular Formula: C9H7NO Molecular Weight (g/mol): 145.16 Synonym: 1,4-Dihydro-4-quinolinone,1H-4-Oxoquinoline,1H-Quinolin-4-one,4-Quinolone,NSC 263800,γ-Quinolone IUPAC Name: 1H-quinolin-4-one SMILES: O=C1C=CNc2ccccc12