accessibility menu, dialog, popup

UserName

CAS RN 5239-82-7

HOO

CAS RN 5239-82-7

IUPAC Name: N-(1,3-dimethyl-1,3,2-diazaborolidin-2-yl)-S-[(1,3-dimethyl-1,3,2-diazaborolidin-2-yl)imino]-λ⁴-sulfanimine
Synonyms: N-(1,3-dimethyl-1,3,2-diazaborolidin-2-yl)-S-[(1,3-dimethyl-1,3,2-diazaborolidin-2-yl)imino]-λ⁴-sulfanimine
Molecular Weight (g/mol): 253.97
Molecular Formula: C8H20B2N6S
InChi Key: BXXOHRWKRXBZTG-UHFFFAOYSA-N
SMILES: CN1CCN(C)B1N=S=NB1N(C)CCN1C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)