accessibility menu, dialog, popup

UserName

CAS RN 5238-27-7

IUPAC Name: (3Z)-4-hydroxy-4-(4-methylphenyl)-2-oxo-N-(1,3-thiazol-2-yl)but-3-enamide
Synonyms: (3Z)-4-hydroxy-4-(4-methylphenyl)-2-oxo-N-(1,3-thiazol-2-yl)but-3-enamide
Molecular Weight (g/mol): 288.32
Molecular Formula: C14H12N2O3S
InChi Key: NADADZQYJBFZGI-FLIBITNWSA-N
SMILES: CC1=CC=C(C=C1)C(\O)=C\C(=O)C(=O)NC1=NC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results