CAS RN 52304-36-6
CAS RN 52304-36-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ethyl 3-(N-Butylacetamido)propanoate, TRC
CAS: 52304-36-6 Molecular Formula: C11 H21 N O3 Molecular Weight (g/mol): 215.29 Synonym: 3-(Butylacetylamino)propionic acid ethyl ester,3-[(N-Butyl-N-acetyl)amino]propionic acid ethyl ester,AI 3-70763,BAAPE,EUS 26-15,Ethyl 3-(N-butylacetamido)propionate,IR 3535,Merck 3535,Quwenzhi,Repellent 3535 IUPAC Name: ethyl 3-[acetyl(butyl)amino]propanoate SMILES: CCCCN(CCC(=O)OCC)C(=O)C