accessibility menu, dialog, popup

UserName

CAS RN 51830-50-3

CH3OSH3COCH3

CAS RN 51830-50-3

IUPAC Name: 1-[(2,2-diethoxyethyl)sulfanyl]-4-methylbenzene
Synonyms: 1-[(2,2-diethoxyethyl)sulfanyl]-4-methylbenzene
Molecular Weight (g/mol): 240.36
Molecular Formula: C13H20O2S
InChi Key: CZOGWUVOCUTQOV-UHFFFAOYSA-N
SMILES: CCOC(CSC1=CC=C(C)C=C1)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)