CAS RN 5178-52-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Isopropyldopamine Hydrochloride, TRC
CAS: 5178-52-9 Molecular Formula: C11H18ClNO2 Molecular Weight (g/mol): 231.72 Synonym: 4-[2-[(1-Methylethyl)amino]ethyl]-1,2-benzenediol Hydrochloride,4-[2-(Isopropylamino)ethyl]-pyrocatechol, Hydrochloride,4-[2-(Isopropylamino)ethyl]pyrocatechol Hydrochloride,Desoxyisoprenaline Hydrochloride IUPAC Name: 4-[2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride SMILES: OC1=C(O)C=CC(CCNC(C)C)=C1.Cl