CAS RN 5157-57-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-dichloro-1-(4-chlorophenyl)ethanone, TRC
CAS: 5157-57-3 Molecular Formula: C8H5OCl3 Molecular Weight (g/mol): 223.48 IUPAC Name: 2,2-dichloro-1-(4-chlorophenyl)ethan-1-one SMILES: O=C(C1=CC=C(Cl)C=C1)C(Cl)Cl