CAS RN 5157-08-4
IUPAC Name:
6-methyl-2,3,4,5-tetrahydropyridazin-3-one
Synonyms:
6-methyl-4,5-dihydro-2H-pyridazin-3-one
Molecular Weight (g/mol):
112.13
Molecular Formula:
C5H8N2O
InChi Key:
VOTFXESXPPEARL-UHFFFAOYSA-N
SMILES:
CC1=NNC(=O)CC1
1
–
1
of
1
results
1
4,5-Dihydro-6-methylpyridazin-3(2H)-one monohydrate, 99%, Thermo Scientific™
CAS: 5157-08-4 MDL Number: MFCD00006455