accessibility menu, dialog, popup

UserName

ONOOH

CAS RN 51362-38-0

IUPAC Name: 6-phenoxypyridine-3-carboxylic acid
Synonyms: 6-phenoxypyridine-3-carboxylic acid
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: GFEUNYLQJDQNAN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results