accessibility menu, dialog, popup

UserName

BrHOOP

CAS RN 51114-94-4

IUPAC Name: (2-carboxyethyl)triphenylphosphanium bromide
Synonyms: (2-carboxyethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 415.27
Molecular Formula: C21H20BrO2P
InChi Key: BVKRDNIULHRLCO-UHFFFAOYSA-N
SMILES: [Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results