accessibility menu, dialog, popup

UserName

CAS RN 5100-98-1

IUPAC Name: 1-chloro-2-iodo-3-methylbenzene
Synonyms: 1-chloro-2-iodo-3-methylbenzene
Molecular Weight (g/mol): 252.48
Molecular Formula: C7H6ClI
InChi Key: FTGLKPMFTLNUBN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1I
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results