CAS RN 50924-49-7
IUPAC Name:
1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxy-1H-imidazole-4-carboxamide
Synonyms:
mizoribine
Molecular Weight (g/mol):
259.22
Molecular Formula:
C9H13N3O6
InChi Key:
HZQDCMWJEBCWBR-UHFFFAOYNA-N
SMILES:
NC(=O)C1=C(O)N(C=N1)C1OC(CO)C(O)C1O
1
–
2
of
2
results
1
Mizoribine, TRC
CAS: 50924-49-7 Molecular Formula: C9 H13 N3 O6 Molecular Weight (g/mol): 259.22 IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxyimidazole-4-carboxamide SMILES: NC(=O)c1ncn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c1O
2
Mizoribine, MedChemExpress
MedChemExpress Mizoribine (NSC 289637), an imidazole nucleoside, inhibits HCV RNA replication with IC50 of approximately 100 μM for anti-HCV activity. Immunosuppressant. Mizoribine, an IMPDH inhibitor, inhibits replication of SARS-CoV with IC50s of 3.5 μg/mL and 16 μg/mL for SARS-CoV Frankfurt-1 and SARS-CoV HKU39849, respectively.