accessibility menu, dialog, popup

UserName

CAS RN 5060-82-2

IUPAC Name: 7-methoxynaphthalen-2-ol
Synonyms: 7-methoxynaphthalen-2-ol
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: UNFNRIIETORURP-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC(O)=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)