CAS RN 50499-60-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(+)-Clenbuterol, TRC
CAS: 50499-60-0 Molecular Formula: C12 H18 Cl2 N2 O Molecular Weight (g/mol): 277.19 Synonym: Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]-, (αS)-,Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]-, (S)-,(αS)-4-Amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]benzenemethanol,(+)-Clenbuterol,(S)-Clenbuterol IUPAC Name: (1S)-1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol SMILES: CC(C)(C)NC[C@@H](O)c1cc(Cl)c(N)c(Cl)c1