CAS RN 4984-22-9
IUPAC Name:
1-methoxypropan-2-ol
Synonyms:
methoxyisopropanol
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
ARXJGSRGQADJSQ-UHFFFAOYNA-N
SMILES:
COCC(C)O
1
–
1
of
1
results
1
(R)-(-)-1-Methoxy-2-propanol, 98+%
CAS: 4984-22-9 MDL Number: MFCD01632587 InChI Key: ARXJGSRGQADJSQ-SCSAIBSYSA-N Synonym: r---1-methoxy-2-propanol,2r-1-methoxypropan-2-ol,r-1-methoxypropan-2-ol,r-1-methoxy-2-propanol,r-1-methoxy-propan-2-ol,2-propanol, 1-methoxy-, 2r,20s-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxahenicosane-1,20-diol,pubchem6714,r-methoxy-propan-2-ol PubChem CID: 2733589 IUPAC Name: (2R)-1-methoxypropan-2-ol SMILES: CC(COC)O