CAS RN 4981-92-4
CAS RN 4981-92-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Benzylbenzimidazole, 97%, Thermo Scientific™
CAS: 4981-92-4 Molecular Formula: C14H21NO2S Molecular Weight (g/mol): 267.387 MDL Number: MFCD00458798 InChI Key: PYBYEBRVJRDWPB-LBPRGKRZSA-N Synonym: s-2-benzyl-2-n-bocamino-ethyl thiol,s-tert-butyl 1-mercapto-3-phenylpropan-2-yl carbamate,tert-butyl n-2s-1-phenyl-3-sulfanylpropan-2-yl carbamate,s-2-benzyl-2-n-bocaminoethyl thiol,s-2-benzyl-2-n-boc-aminoethyl thiol,s-2-n-boc-2-amino-3-phenylpropane-1-thiol,s-1-mercaptomethyl-2-phenylethyl carbamic acid tert-butyl ester PubChem CID: 15003862 IUPAC Name: tert-butyl N-[(2S)-1-phenyl-3-sulfanylpropan-2-yl]carbamate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CS