accessibility menu, dialog, popup

UserName

CAS RN 497959-31-6

OFFFHOO

CAS RN 497959-31-6

IUPAC Name: 2-hydroxy-3-(trifluoromethoxy)benzaldehyde
Synonyms: 2-hydroxy-3-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: QKIITUNJEPEAIC-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results