CAS RN 4959-60-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,5-Dimethoxy-2-nitrobenzamide, TRC
CAS: 4959-60-8 Molecular Formula: C9H10N2O5 Molecular Weight (g/mol): 226.19 Synonym: Veratramide, 6-nitro- (7CI,8CI),4,5-Dimethoxy-2-nitrobenzamide IUPAC Name: 4,5-dimethoxy-2-nitrobenzamide SMILES: COc1cc(C(=O)N)c(cc1OC)[N+](=O)[O-]