accessibility menu, dialog, popup

UserName

OOHO

CAS RN 4958-02-5

IUPAC Name: 3-(benzyloxy)cyclobutane-1-carboxylic acid
Synonyms: 3-(benzyloxy)cyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: YNNOFVDQHAHVFG-UHFFFAOYSA-N
SMILES: OC(=O)C1CC(C1)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results