CAS RN 488841-24-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy Estrone 1-N3-Adenine, TRC
CAS: 488841-24-3 Molecular Formula: C23H25N5O3 Molecular Weight (g/mol): 419.48 Synonym: 1-(6-Amino-3H-purin-3-yl)-3,4-dihydroxyestra-1,3,5(10)-trien-17-one,4-OH-E1-1-N3Ade SMILES: C[C@]12CC[C@H]3[C@H]([C@@H]1CCC2=O)CCC4=C3C(=CC(=C4O)O)N5C=NC(=C6C5=NC=N6)N