CAS RN 487-03-6
IUPAC Name:
1-methyl-2H,7H,9H-pyrido[3,4-b]indol-7-one
Synonyms:
harmol
Molecular Weight (g/mol):
198.23
Molecular Formula:
C12H10N2O
InChi Key:
LBBJNGFCXDOYMQ-UHFFFAOYSA-N
SMILES:
CC1=C2NC3=CC(=O)C=CC3=C2C=CN1
1
–
1
of
1
results
1
Harmol, TRC
CAS: 487-03-6 Molecular Formula: C12 H10 N2 O Molecular Weight (g/mol): 198.22 Synonym: 9H-Pyrido[3,4-b]indol-7-ol, 1-methyl-,Harmol (6CI,7CI),NSC 72292,1-methyl-9H-pyrido[3,4-b]indol-7-ol IUPAC Name: 1-methyl-9H-pyrido[3,4-b]indol-7-ol SMILES: Cc1nccc2c3ccc(O)cc3[nH]c12