accessibility menu, dialog, popup

UserName

CAS RN 4790-79-8

CH3OOOO

CAS RN 4790-79-8

IUPAC Name: 7-methoxy-1-benzofuran-2-carboxylate
Synonyms: 7-methoxy-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 191.16
Molecular Formula: C10H7O4
InChi Key: UOCNTRAAJNWDND-UHFFFAOYSA-M
SMILES: COC1=C2OC(=CC2=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results