accessibility menu, dialog, popup

UserName

H3COOFFFBr

CAS RN 478484-53-6

IUPAC Name: 2-(bromomethyl)-1-methoxy-4-(trifluoromethoxy)benzene
Synonyms: 2-(bromomethyl)-1-methoxy-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 285.06
Molecular Formula: C9H8BrF3O2
InChi Key: HSHYZUINFUNPJO-UHFFFAOYSA-N
SMILES: COC1=C(CBr)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results