CAS RN 477860-13-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Fluoro-4-phenylphenol, TRC
CAS: 477860-13-2 Molecular Formula: C12 H9 F O Molecular Weight (g/mol): 188.2 Synonym: 2-Fluoro[1,1'-biphenyl]-4-ol IUPAC Name: 3-fluoro-4-phenyl-phenol SMILES: Oc1ccc(c(F)c1)c2ccccc2