accessibility menu, dialog, popup

UserName

CH3HNO

CAS RN 4753-75-7

IUPAC Name: [(furan-2-yl)methyl](methyl)amine
Synonyms: 2-furanmethanamine, N-methyl-
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: DGLIOWSKNOCHEX-UHFFFAOYSA-N
SMILES: CNCC1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results