CAS RN 47363-37-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Miconazole Impurity I, TRC
CAS: 47363-37-1 Molecular Formula: C18 H15 Cl3 N2 O Molecular Weight (g/mol): 381.683 Synonym: 1-[2-[(2-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-Imidazole,Miconazole EP Impurity I IUPAC Name: 1-[2-[(2-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole SMILES: Clc1ccc(C(Cn2ccnc2)OCc3ccccc3Cl)c(Cl)c1