accessibility menu, dialog, popup

UserName

CAS RN 4724-89-4

CH3H3CCH3H3C

CAS RN 4724-89-4

IUPAC Name: 1,3,5,5-tetramethylcyclohexa-1,3-diene
Synonyms: 1,3,5,5-tetramethylcyclohexa-1,3-diene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: SZHAWDAGEJWQJK-UHFFFAOYSA-N
SMILES: CC1=CC(C)=CC(C)(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results