CAS RN 4663-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyridine-3,5-dicarboxamide, TRC
CAS: 4663-99-4 Molecular Formula: C7H7N3O2 Molecular Weight (g/mol): 165.14 IUPAC Name: pyridine-3,5-dicarboxamide SMILES: NC(=O)c1cncc(c1)C(=O)N