accessibility menu, dialog, popup

UserName

OOH

CAS RN 4541-14-4

IUPAC Name: 4-(benzyloxy)butan-1-ol
Synonyms: 4-(benzyloxy)butan-1-ol
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: TYROJDFHUXSBHC-UHFFFAOYSA-N
SMILES: OCCCCOCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results